0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 95%
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 95%
:
Apollo Scientific
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 95%
:
Apollo Scientific
:
146504-07-6
:
:
C₁₁H₂₂N₂O₂
:
MFCD08726023
:
214.31 g/mol
:
GHS07
:
Refrigerator