Order Entry
Sweden
ContactUsLinkComponent
1-Methylpyrrole 98%
1-Methylpyrrole 98%
Catalog # APOSOR5110-100G
Supplier:  Apollo Scientific
CAS Number:  
1-Methylpyrrole 98%
Catalog # APOSOR5110-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :81.12 g/mol
Boiling Pt:112…113 °C
Melting Pt:–57 °C
Density:0.91 g/cm³ (25 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
CAS Number:96-54-8
Formula:C₅H₇N
MDL Number:MFCD00005345