Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Cyanobenzaldehyde
4-Cyanobenzaldehyde 98%
4-Cyanobenzaldehyde 98%
:
Apollo Scientific
4-Cyanobenzaldehyde 98%
:
Apollo Scientific
:
105-07-7
:
:
C₈H₅NO
:
MFCD00003376
:
131.13 g/mol
:
133 °C/12mmHg
:
98…100 °C
:
Ambient
:
203-267-3
:
GHS07