Fmoc-2-amino-5-chlorobenzoic acid
Catalog # APOSOR470395-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:393.83 g/mol
Storage Temperature:Ambient
CAS Number:332121-90-1
Formula:C₂₂H₁₆ClNO₄
MDL Number:MFCD02682159