(S)-1-Methyl-3-pyrrolidinol 97%
Catalog # APOSOR4625-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:101.15 g/mol
Storage Temperature:Ambient
CAS Number:104641-59-0
Formula:C₅H₁₁NO
MDL Number:MFCD03788748