Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-methoxyphenol
4-Chloro-2-methoxyphenol 98%
4-Chloro-2-methoxyphenol 98%
APOSOR4506-1G
:
Apollo Scientific
:
4-Chloro-2-methoxyphenol 98%
APOSOR4506-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
158.58 g/mol
:
241 °C
:
16…17 °C
:
1.304 g/cm³ (25 °C)
:
113 °C
:
GHS07
:
Ambient
:
16766-30-6
:
C₇H₇ClO₂
:
MFCD00070774