Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzyl-4-oxoazepane-1-carboxylate
Benzyl-4-oxoazepane-1-carboxylate
Benzyl-4-oxoazepane-1-carboxylate
Irritant
Benzyl-4-oxoazepane-1-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Benzyl-4-oxoazepane-1-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
83621-33-4
:
Formula:
C₁₄H₁₇NO₃
MDL Number:
MFCD03788442
:
247.2897 g/mol
:
GHS07
Storage Temperature:
Ambient