Gå till Rubrik
Gå till Huvudinnehåll
Gå till Sidfot
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(-)-3-Amino-1,2-propanediol
(S)-(-)-3-Amino-1,2-propanediol 97%
(S)-(-)-3-Amino-1,2-propanediol 97%
(S)-(-)-3-Amino-1,2-propanediol 97%
APOSOR42150-100G
:
Apollo Scientific
:
(S)-(-)-3-Amino-1,2-propanediol 97%
APOSOR42150-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
91.11 g/mol
:
117…119 °C/0.4mmHg
:
55 °C
:
1.175 g/cm³ (25 °C)
:
Ambient
:
61278-21-5
:
C₃H₉NO₂
:
MFCD00798260