
Ethyl-7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR40503-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:224.65 g/mol
:GHS07
Storage Temperature:Omgivande
CAS Number:1204501-38-1
Formula:C₁₀H₉BrN₂O₂
MDL Number:MFCD11505035