Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-3-methoxybenzoic acid
4-Chloro-3-methoxybenzoic acid
4-Chloro-3-methoxybenzoic acid
4-Chloro-3-methoxybenzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloro-3-methoxybenzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
85740-98-3
:
Formula:
C₈H₇ClO₃
MDL Number:
MFCD00269640
:
186.5900 g/mol
Storage Temperature:
Ambient