Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
Irritant
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
Supplier:
Apollo Scientific
About this item
CAS Number:
15883-20-2
:
Formula:
C₁₄H₂₀N₂O
MDL Number:
MFCD01701244
:
232.32 g/mol
Melting Pt:
114…117 °C
:
GHS07
Storage Temperature:
Ambient