Order Entry
Sweden
ContactUsLinkComponent
H-Cha-OMe·HCl 95%
H-Cha-OMe·HCl 95%
Catalog # APOSOR32767-1G
Supplier:  Apollo Scientific
CAS Number:  
H-Cha-OMe·HCl 95%
Catalog # APOSOR32767-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Förpackningstyp:
    Bottle
  • Förp.:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.7 g/mol
Storage Temperature:Omgivande
CAS Number:144600-01-1
Formula:C₁₀H₁₉NO₂·HCl
Storage Temperature:Ambient