(2-Ethylhexyl)magnesium bromide 1 M in diethyl ether
Catalog # APOSOR320020-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:217.43 g/mol
Storage Temperature:Ambient
CAS Number:90224-21-8
Formula:C₈H₁₇BrMg
MDL Number:MFCD02260220