Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-[(Trimethylsilyl)ethynyl]benzonitrile
3-[(Trimethylsilyl)ethynyl]benzonitrile 99%
3-[(Trimethylsilyl)ethynyl]benzonitrile 99%
3-[(Trimethylsilyl)ethynyl]benzonitrile 99%
Supplier:
Apollo Scientific
Certificates
Expand image
3-[(Trimethylsilyl)ethynyl]benzonitrile 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
190771-22-3
:
Formula:
C₁₂H₁₃NSi
MDL Number:
MFCD09260494
:
199.33 g/mol
Melting Pt:
42…47 °C
:
GHS07
Storage Temperature:
Ambient