Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octachloronaphthalene
Octachloronaphthalene, Technical Grade
Octachloronaphthalene, Technical Grade
Irritant
Octachloronaphthalene, Technical Grade
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Octachloronaphthalene, Technical Grade
Supplier:
Apollo Scientific
About this item
CAS Number:
2234-13-1
:
Formula:
C₁₀Cl₈
:
403.73 g/mol
:
GHS07
Storage Temperature:
Ambient