Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Aminoisoindolin-1-one
5-Aminoisoindolin-1-one
5-Aminoisoindolin-1-one
Irritant
5-Aminoisoindolin-1-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Aminoisoindolin-1-one
Supplier:
Apollo Scientific
About this item
CAS Number:
222036-66-0
:
Formula:
C₈H₈N₂O
MDL Number:
MFCD10000828
:
148.1600 g/mol
:
GHS07
Storage Temperature:
Ambient