
(R)-(-)-1-Aminopropan-2-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2799-16-8
:
Formula:C₃H₉NO
MDL Number:MFCD00064428
:75.11 g/mol
Boiling Pt:160 °C
Melting Pt:24…26 °C
Density:0.954 g/cm³ (25 °C)
Flash Pt:71 °C
Storage Temperature:Ambient
EINECS:220-532-9
UN:3259
ADR:8,II
:GHS05