3-Benzyloxy-1-propanol 98%
Catalog # APOSOR301193-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:166.22 g/mol
Boiling Pt:111…114 °C/2mmHg
Density:1.049 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:4799-68-2
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00029659