Order Entry
Sweden
ContactUsLinkComponent
3-(Aminomethyl)benzonitrile 98%
Corrosive
Irritant
3-(Aminomethyl)benzonitrile 98%
Catalog # APOSOR301027-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-(Aminomethyl)benzonitrile 98%
Catalog # APOSOR301027-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :132.1600 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:10406-24-3
Formula:C₈H₈N₂
MDL Number:MFCD06797832