Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromobenzoyl chloride
2-Bromobenzoyl chloride
2-Bromobenzoyl chloride
:
Apollo Scientific
2-Bromobenzoyl chloride
:
Apollo Scientific
:
7154-66-7
:
:
C₇H₄BrClO
:
MFCD00000655
:
219.46 g/mol
:
245 °C
:
44477 °C
:
1.679 g/cm³ (25 °C)
:
113 °C
:
Refrigerator
:
230-507-4
:
3265
:
8,II
:
GHS05