Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-N-ethylglycine
N-Boc-N-ethylglycine
N-Boc-N-ethylglycine
Irritant
N-Boc-N-ethylglycine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-Boc-N-ethylglycine
Supplier:
Apollo Scientific
About this item
CAS Number:
149794-10-5
:
Formula:
C₉H₁₇NO₄
MDL Number:
MFCD00672508
Melting Pt:
87…88 °C
:
203.24 g/mol
:
GHS07
Storage Temperature:
Omgivande