
(R)-2-Acetoxy-2-phenylacetic acid 98%
Catalog # APOSOR18839-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:97…99 °C
:194.19 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:51019-43-3
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004249