0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Methyl 4-chlorobenzo[b]thiophene-2-carboxylate
Methyl-4-chlorobenzo[b]thiophene-2-carboxylate
Methyl-4-chlorobenzo[b]thiophene-2-carboxylate
:
Apollo Scientific
Methyl-4-chlorobenzo[b]thiophene-2-carboxylate
:
Apollo Scientific
:
35212-95-4
:
:
C₁₀H₇ClO₂S
:
MFCD07371546
:
95…97 °C
:
GHS07
:
Ambient