Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-bromopyrazine
2-Amino-5-bromopyrazine
2-Amino-5-bromopyrazine
Irritant
2-Amino-5-bromopyrazine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-5-bromopyrazine
Supplier:
Apollo Scientific
About this item
CAS Number:
59489-71-3
:
Formula:
C₄H₄BrN₃
MDL Number:
MFCD00235015
:
174.00 g/mol
Melting Pt:
113…117 °C
:
GHS07
Storage Temperature:
Ambient