

3-Bromopyruvic acid hydrate 95%
Catalog # APOSOR10838-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:79…82 °C
Storage Temperature:Kylskåp
UN:3261
ADR:8,III
:166.96 g/mol (anhy)
:GHS05
:GHS07
CAS Number:206860-50-6
:1113-59-3
Formula:C₃H₃BrO₃.xH₂O
MDL Number:MFCD00149107