


Chlorotrimethylsilane 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75-77-4
:
Formula:C₃H₉ClSi
MDL Number:MFCD00000502
Boiling Pt:56…58 °C
Melting Pt:∼ …58 °C
Density:0.856 g/cm³ (25 °C)
Flash Pt:–18 °C
Storage Temperature:Refrigerator
EINECS:200-900-5
ADR:3+8,II
:108.64 g/mol
UN:1298
:GHS02
:GHS05
:GHS07