
(R)-(-)-2-Amino-1-propanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35320-23-1
:
Formula:C₃H₉NO
MDL Number:MFCD00064413
:75.11 g/mol
Boiling Pt:168…176 °C
Density:0.965 g/cm³ (25 °C)
Flash Pt:83 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05