D-α-Phenylalanine 97%
Catalog # APOSOR0702-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:165.19 g/mol
Melting Pt:273…276 °C
EINECS:211-603-5
:GHS07
Storage Temperature:Ambient
CAS Number:673-06-3
Formula:C₉H₁₁NO₂
MDL Number:MFCD00004270