Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-4-hydroxybenzonitrile
3-Amino-4-hydroxybenzonitrile
3-Amino-4-hydroxybenzonitrile
3-Amino-4-hydroxybenzonitrile
Leverantör:
Apollo Scientific
Certifikat
Expand image
3-Amino-4-hydroxybenzonitrile
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
14543-43-2
:
Formel:
C₇H₆N₂O
MDL-nummer:
MFCD00234273
:
134.14 g/mol
Smält Pt:
151…153 °C
:
GHS07
Förvaringstemperatur:
Ambient