Order Entry
Sweden
ContactUsLinkComponent
Potassium hydroxide 99.995%
Potassium hydroxide 99.995%
Catalog # APOSIN2917-100G
Supplier:  Apollo Scientific
CAS Number:  
Potassium hydroxide 99.995%
Catalog # APOSIN2917-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :56.11 g/mol
Boiling Pt:1320…1324 °C
Melting Pt:360…361 °C
Density:2.044 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553