Order Entry
Sweden
ContactUsLinkComponent
Copper(II) oxide 99,9%
Copper(II) oxide 99,9%
Catalog # APOSIN1576-500G
Supplier:  Apollo Scientific
CAS Number:  
Copper(II) oxide 99,9%
Catalog # APOSIN1576-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :79.55 g/mol
Melting Pt:1362 °C
Density:6.315 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:1317-38-0
Formula:CuO
MDL Number:MFCD00010979