D-(+)-Mannose 99,5%
Supplier: Apollo Scientific
Jump to:
About this item
- Heavy metals: <10ppm
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Melting Pt:133…140 °C
- Density:1.54 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:222-392-4
3 Options Available Below
Product Details & Documents
- Heavy metals: <10ppm
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Melting Pt:133…140 °C
- Density:1.54 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:222-392-4



