Glycine ≥99%
Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.
- Ph. Eur.
- Pyrogen free
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :240…245 °C (dec.)
- :1.595 g/cm³ (25 °C)
- :Ambient
- :200-272-2
Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.
- Ph. Eur.
- Pyrogen free
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :240…245 °C (dec.)
- :1.595 g/cm³ (25 °C)
- :Ambient
- :200-272-2
Product Family Options
- Pack typePk