Order Entry
Sweden
Orders LinkContactUsLinkComponent
Fusidate sodium

Fusidate sodium

Irritant
Supplier:   Apollo Scientific
Fusidate sodium
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

Suppresses nitric oxide lysis of pancreatic islet cells.

Synonyms:  Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
  • CAS Number:751-94-0
  • Formula:C₃₁H₄₇NaO₆
  • MDL Number:MFCD09054714
  • Molecular Weight:538.701 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 875,00 kr
    1 875,00 kr
    1 * 1 g
       1 875,00 kr

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 3 750,00 kr
    3 750,00 kr
    1 * 5 g
       3 750,00 kr

  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 5 975,00 kr
    5 975,00 kr
    1 * 10 g
       5 975,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.