Order Entry
Sweden
ContactUsLinkComponent
Calcium chloride dihydrate
Calcium chloride dihydrate
Catalog # APOSBIA4689-250G
Supplier:  Apollo Scientific
CAS Number:  
Calcium chloride dihydrate
Catalog # APOSBIA4689-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :147.01 g/mol
Melting Pt:175 °C
Density:1.85 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613