Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Proline
D-(+)-Proline
D-(+)-Proline
D-(+)-Proline
Supplier:
Apollo Scientific
Certificates
Expand image
D-(+)-Proline
Supplier:
Apollo Scientific
About this item
CAS Number:
344-25-2
:
Formula:
C₅H₉NO₂
MDL Number:
MFCD00064317
:
115.13 g/mol
Melting Pt:
223 °C (dec.)
EINECS:
206-452-7
:
GHS07
Storage Temperature:
Ambient