Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tricine
Tricine ≥99%
Tricine ≥99%
Irritant
Tricine ≥99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Tricine ≥99%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
5704-04-1
:
Formula:
(HOCH₂)₃CNHCH₂CO₂H
MDL Number:
MFCD00004277
:
179.17 g/mol
Melting Pt:
187 °C (dec.)
EINECS:
227-193-6
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation