Human recombinant mesothelin (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,01 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human Mesothelin, His Tag, low endotoxin at 0,5 μg/ml (100 μl/well) can bind Anti-MSLN MAb, Human IgG1 with a linear range of 0,2 - 3 ng/ml (QC tested)
- Gene ID:Q13421
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:33,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT