Human recombinant IL-29 (from HEK293)
Catalog # ACRBIL9-H4228-20UG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pk:20 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:Q8IU54
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IFNL1|IL29|IFNλ1|IFN lambda 1|Interleukin-29
- Protein/peptide name:IL-29
- Purity:>95%
- Molecular weight:21,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT