Order Entry
Sweden
ContactUsLinkComponent
 
Human recombinant IL-12B and IL-12A (from HEK293)
  ACRBIL2-H8210-25UG
 :  
Human recombinant IL-12B and IL-12A (from HEK293)
  ACRBIL2-H8210-25UG
 :  

 

  • Pk:
    25 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilized Human IL-12B and IL-12A heterodimer protein, His Tag and Flag Tag (Cat. No. ACRBIL2-H4210) at 0,5 μg/ml (100 μl/well) can bind anti-IL23A and IL12B MAb (P40 domain) with a linear range of 0,2 - 2 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised biotinylated human IL-12 R beta 1, Fc,Avitag (Cat. No. ACRBILB-H82F7) at 1 μg/ml (100 μl/well) on streptavidin (Cat. No. ACRBSTN-N5116) precoated (0,5 μg/well) plate, can bind human IL-12B and IL-12A heterodimer protein, His Tag and Flag Tag (Cat. No. ACRBIL2-H4210) with a linear range of 0,039 - 0,625 μg/ml (routinely tested). Human IL-12 R beta 1, Fc Tag (Cat. No. ACRBILB-H5255) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human IL-12B and IL-12A heterodimer protein, His tag and flag tag (Cat. No. ACRBIL2-H4210) with an affinity constant of 2,06 nM as determined in a SPR assay (Biacore T200) (routinely tested).; Loaded Anti-IL23A and IL12B MAb (P40 domain) on protein A biosensor, can bind Human IL-12B and IL-12A heterodimer protein, His tag and flag tag (Cat. No. ACRBIL2-H4210) with an affinity constant of 1,07 nM as determined in BLI assay (ForteBio Octet Red96e) (routinely tested). Loaded human IL-12B and IL-12A heterodimer protein, His Tag and Flag Tag (Cat. No. ACRBIL2-H4210) on HIS1K Biosensor, can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with an affinity constant of 92.2 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Human IL-12B and IL-12A Heterodimer Protein, His Tag and Flag Tag (Cat. No. ACRBIL2-H4210) stimulates the proliferation of PHA-P activated PBMC. The ED50 for this effect is 0,05 ng/ml (routinely tested).
  • Gene ID:
    P29460-1 (IL12B) and P29459-1 (IL12A)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IL12|p70|Interleukin-12
  • Protein/peptide name:
    IL-12B and IL-12A
  • Purity:
    >90%
  • Molecular weight:
    36,6 kDa (IL12B) and 23,8 kDa (IL12A)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT