Order Entry
Sweden
ContactUsLinkComponent
Human recombinant CLEC12A (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Human recombinant CLEC12A (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant CLEC12A (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 ℃, aviod light
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    2e5 of Anti-CLEC12A CAR-293 cells were stained with 100 μl of 10 μg/ml of FITC-Labeled Human CLEC12A, His Tag (Cat. No. CLA-HF247) and negative control protein respectively. FITC signal was used to evaluate the binding activity (QC tested).
  • Gene ID:
    Q5QGZ9-2
  • Protein synonyms:
    CLL-1|MICL|CLEC12A|CD371|CLL1|DCAL-2|DCAL2
  • Protein/peptide name:
    CLEC12A
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    25,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications