Order Entry
Sweden
ContactUsLinkComponent
Human recombinant TNFSF11 (from HEK293 cells)
Human recombinant TNFSF11 (from HEK293 cells)
Catalog # ACRBRAL-H5240-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant TNFSF11 (from HEK293 cells)
Catalog # ACRBRAL-H5240-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (active trimer) (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Anti-TNFSF11 (human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human TNFSF11, His Tag (active trimer) (ACRBRAL-H5240) with an affinity constant of 0,274 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Captured Human RANK, Mouse IgG2a Fc Tag, low endotoxin (ACRBRAK-H5251) on CM5 chip via Anti-Mouse antibodies surface can bind Human TNFSF11, His Tag (active trimer) (ACRBRAL-H5240) with an affinity constant of 0,309 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Human RANK, Mouse IgG2a Fc Tag, low endotoxin (ACRBRAK-H5251) on AMC Biosensor, can bind Human TNFSF11, His Tag (active trimer) (ACRBRAL-H5240) with an affinity constant of 0.409 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Anti-TNFSF11 MAb (human IgG1) on AHC Biosensor, can bind Human TNFSF11, His Tag (active trimer) (ACRBRAL-H5240) with an affinity constant of 0.239 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    AAC51762.1
  • Protein synonyms:
    OPGL|ODF|TRANCE|CD254|RANKL
  • Protein/peptide name:
    TNFSF11
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    22,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Human TNFSF11 / RANKL / CD254 Protein, His Tag (active trimer) (MALS verified).