Order Entry
Sweden
ContactUsLinkComponent
MERS recombinant nucleocapsid protein (from HEK293 cells)
MERS recombinant nucleocapsid protein (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
MERS recombinant nucleocapsid protein (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    MERS
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    K0BVN3-1
  • Protein synonyms:
    NP|Nucleocapsid protein|Protein N|COVID-19
  • Protein/peptide name:
    Nucleocapsid protein
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    46,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 . Normally Trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications