Human recombinant CBLB (from E. coli), Acro Biosystems
Human CBLB/RNF56 protein, His tag.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:Q13191-1
- Protein synonyms:RNF56|CBLB|CBL-B
- Protein/peptide name:CBLB
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:47,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 40 mM HEPES, 120 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT