Human recombinant CD27 ligand (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD27 (Cat. No. CD7-H522b) at 2 μg/ml (100 μl/well) can bind Human CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525a) with a linear range of 0,2 - 6 ng/ml (QC tested).
- Gene ID:P32970-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
- Protein/peptide name:CD27 Ligand
- Purity:>85%
- Molecular weight:44,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT