Human recombinant LILRB2 (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
- Gene ID:Q8N423
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD85d|ILT4|LILRB2|LIR2|MIR10
- Protein/peptide name:LILRB2
- Purity:>90%
- Molecular weight:76,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together