1-Chloro-2,4,6-trifluorobenzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2106-40-3
:
Formula:C₆H₂ClF₃
MDL Number:MFCD00012239
:166.528 g/mol
Boiling Pt:124 °C
Melting Pt:–1 °C
Flash Pt:41 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07