Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-chloro-5-fluoropyridine
2-Bromo-3-chloro-5-fluoropyridine
2-Bromo-3-chloro-5-fluoropyridine
Irriterande
2-Bromo-3-chloro-5-fluoropyridine
Leverantör:
Apollo Scientific
Certifikat
Expand image
Irriterande
2-Bromo-3-chloro-5-fluoropyridine
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
1214326-94-9
:
Formel:
C₅H₂BrClFN
MDL-nummer:
MFCD13185350
:
210.431 g/mol
:
GHS07
Förvaringstemperatur:
Ambient