(4'-Fluoro-1,1'-biphenyl-4-yl)methanol
Catalog # APOSPC300617-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.224 g/mol
Storage Temperature:Ambient
CAS Number:147497-56-1
Formula:C₁₃H₁₁FO
MDL Number:MFCD01862513