Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-1H-indole-2-carbonitrile
4-Chloro-1H-indole-2-carbonitrile 95%
4-Chloro-1H-indole-2-carbonitrile 95%
4-Chloro-1H-indole-2-carbonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloro-1H-indole-2-carbonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
4404-11-9
:
Formula:
C₉H₅ClN₂
MDL Number:
MFCD23135759
:
176.602 g/mol
:
GHS07
Storage Temperature:
Ambient